Learn More
Medchemexpress LLC HY-15335 5mg , Nutlin-3b CAS:675576-97-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY153355MG
Encompass_Preferred
HY-15335 5mg Nutlin-3b CAS: 675576-97-3 Nutlin-3b is a p53/MDM2 inhibitor with an IC50 of 13.6 μM. Nutlin-3b is 150 times less potent in binding to MDM2 than Nutlin-3a. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1483