Learn More
Alkali Scientific Nutrient Broth, 1 Kg
SDP
Supplier:  Alkali Scientific UTR001CP
Encompass_Preferred
Nutrient Broth (1 kg) is a versatile liquid medium used to cultivate a wide range of microorganisms in microbiological studies. It contains essential nutrients such as peptones and sodium chloride, providing the necessary environment for the growth of both bacteria and fungi. The broth is often used in bacterial cultivation, biochemical testing, and in preparing cultures for various microbiological assays. It is ideal for supporting microbial growth in clinical diagnostics, industrial testing, and research applications where liquid media is required for suspensions, dilutions, or testing protocols.
Catalog No. 50-006-60540