Learn More
Cayman Chemical nutrophIl Elasts InhIbItr 50mg
SDP
Supplier:  Cayman Chemical 1492250 MG
Encompass_Preferred
An N-benzoylindazole derivative that selectively targets the binding domain of neutrophil elastase (IC50 = 7 nM)
Catalog No. 50-297-0684