Learn More
Bio X Cell InVivoMAb mouse IgG2a isotype control (clone C1.18.4) - liquid PBS formulation, 1-100 mg sizes
Supplier: Bio X Cell BE0085
InVivoMAb mouse IgG2a isotype control (unknown specificity) is a non-reactive, isotype-matched control antibody designed for use alongside mouse IgG2a primary antibodies in both in vivo and in vitro experiments. Supplied as a liquid in PBS without stabilizers or preservatives, it is commonly used as a negative control in flow cytometry, neutralization, depletion, and immune stimulation studies. Store the antibody at 4°C and do not freeze.
- Clone: C1.18.4.
- Isotype: Mouse IgG2a, κ.
- Formulation: Liquid in PBS, pH 7.0; no stabilizers or preservatives.
- Available sizes: 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, and custom amounts.
- Applications: Flow cytometry, in vivo and in vitro isotype control, neutralization, cell depletion, immune stimulation assays.
- Storage: Store at 4°C; do not freeze.
- Research identifier: RRID AB_1107771.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.