Learn More
Medchemexpress LLC HY-19624 5mg Medchemexpress, NVP-ACC789 CAS:300842-64-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY196245MG
Encompass_Preferred
Medchemexpress, HY-19624 5mg NVP-ACC789 CAS:300842-64-2 NVP-ACC789 is an inhibitor of human VEGFR-1, VEGFR-2 (mouse VEGFR-2), VEGFR-3 and PDGFR-β with IC50s of 0.38, 0.02 (0.23), 0.18, 1.4 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2151