Learn More
Abcam NVP-BEZ235, PI3K and mTOR inhibitor, 5MG
Supplier:  Abcam AB1208825MG
Encompass_Preferred
MW 469.5 Da, Purity >98%. Potent, ATP-competitive Phosphatidylinositol 3-kinase (PI3K) inhibitor (IC₅₀ = 4, 5, 7 and 75 nM for PI3Kα, -γ, -δ, and -β respectively) and mTOR inhibitor. Able to directly induce G₁ arrest and inhibit angiogenesis in vitro and potentiate the efficacy of other anticancer agents in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7174