Learn More
TARGETMOL CHEMICALS INC NVP-BSK805 2HCL 1092499- 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T62945MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NVP-BSK805 2HCl (1092499-93-8(free base)) (BSK 805)(IC50=0.5 nM) a specific and effective ATP-competitive JAK2 inhibitor is more than 20-fold specificity over JAK1 JAK3 and TYK2.
Catalog No. 50-257-9451