Learn More
Medchemexpress LLC Nvp-bsk805 dihydrochloride | 1942919-79-0 | 99.4% | 563.47 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14722A10MM/1ML
Encompass_Preferred
NVP-BSK805 dihydrochloride is a potent, ATP-competitive JAK2 inhibitor. It targets JAK2 JH1, wild-type JAK2, and FL JAK2 V617F activity, effectively suppressing the growth of JAK2V617F-bearing acute myeloid leukemia cell lines and blocking STAT5 phosphorylation. It also enhances the efficacy of vincristine treatment in drug-resistant cancer cells.
  • ATP-competitive inhibitor.
  • Highly potent and selective JAK2 inhibitor.
  • Inhibits full-length wild-type JAK2 and FL JAK2 V617F activity.
  • Suppresses the growth of JAK2V617F-bearing acute myeloid leukemia cell lines.
  • Blocks STAT5 phosphorylation.
  • Shows bias for JAK2 over JAK1 and JAK3 inhibition.
  • Improves P-gp inhibitory activity.
  • Increases sensitization of drug-resistant cancer cells to vincristine treatment.
Catalog No. 50-003-59570