Learn More
Medchemexpress LLC HY-15190 10mg , NVP-HSP990 CAS:934343-74-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1519010MG
Encompass_Preferred
HY-15190 10mg NVP-HSP990 CAS: 934343-74-5 NVP-HSP990 is a potent and selective Hsp90 inhibitor, with IC50 values of 0.6, 0.8, and 8.5 nM for Hsp90α, Hsp90β, and Grp94, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9741