Learn More
Medchemexpress LLC NVP-LCQ195 | 902156-99-4 | C17H19Cl2N5O4S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY15241100MG
Encompass_Preferred
NVP-LCQ195 is an inhibitor of CDK1/2/3/5/9, which induces cell cycle arrest and apoptotic cell death in multiple myeloma (MM) cells. It is supplied as a white to off-white solid and is recommended for research use only.
  • Inhibits CDK1/2/3/5/9
  • Induces cell cycle arrest in multiple myeloma cells
  • Triggers apoptotic cell death in multiple myeloma cells
  • Decreases transcriptional signatures related to oncogenesis, drug resistance, and stem cell renewal in MM cell lines
  • Appears as a white to off-white solid
Catalog No. 50-003-73549