Learn More
Selleck Chemical LLC NVP-TNKS656 5mg 1419949-20-4
SDP
Supplier:  Selleck Chemical LLC S72385MG
Encompass_Preferred
NVP-TNKS656 is a highly potent, selective, and orally active tankyrase inhibitor with IC50 of 6 nM for TNKS2, > 300-fold selectivity against PARP1 and PARP2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4545