Learn More
Cayman Chemical NVP-TNKS656 5mg
SDP
Supplier:  Cayman Chemical 199305 MG
Encompass_Preferred
A TNKS2 inhibitor (IC50 = 6 nM); inhibits Wnt ligand-induced signaling in an HEK293 cell reporter assay (IC50 = 3.5 nM); reduces Axin 2 mRNA expression in MMTV-Wnt1 tumor bearing mice
Catalog No. 50-298-2469