Learn More
Medchemexpress LLC NVS-ZP7-4 10mM 1mL | 2349367-89-9 | 1 ML

Supplier: Medchemexpress LLC HY11439510MM 1ML
NVS-ZP7-4 is a small-molecule inhibitor of the zinc transporter SLC39A7 (ZIP7), supplied as a 10 mM, 1 mL stock solution for cellular and biochemical studies. It modulates ER zinc levels, influences Notch signaling and ferroptosis, and is used as a chemical probe for ZIP7-related research.
- Inhibits zinc transporter SLC39A7 (ZIP7).
- Supplied as a 10 mM stock solution in a 1 mL volume.
- Molecular weight 501.62 g/mol; formula C28H28FN5OS.
- Purity ≥98.0% (HPLC).
- Stable frozen; in solvent store at -80°C for up to 6 months, or at -20°C for up to 1 month; protect from light.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.