Learn More
Selleck Chemical LLC NVS-ZP7-4
SDP
Supplier:  Selleck Chemical LLC S666825MG
Encompass_Preferred
NVS-ZP7-4 a Zinc transporter SLC39A7 (ZIP7) inhibitor is the first reported chemical tool to probe the impact of modulating ER zinc levels and investigate ZIP7 as a novel druggable node in the Notch pathway
Catalog No. 50-000-01849