Learn More
Aviva Systems Biology Corp NXPH4 Antibody - N-terminal region (ARP53543_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53543_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NXPH4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NXPH4. Alternate names: NPH4ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-63040