Learn More
ABclonal Technology NXT2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A13822500UL
Encompass_Preferred
The protein encoded by this gene contains a nuclear transport factor 2 (NTF2) domain, which plays an important role in the trafficking of macromolecules, ions, and small molecules between the cytoplasm and nucleus. This protein may also have a role in mRNA nuclear export. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.
Catalog No. 50-000-99232