Learn More
Medchemexpress LLC NY-ESO-1 (157-165) peptide | 202815-17-6 | 99.75% | 1094.35 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP1049310MG
Encompass_Preferred
NY-ESO-1 (157-165) peptide is a peptide fragment derived from the NY-ESO-1 protein. It has the ability to activate the immune system, particularly in individuals who are HLA-A2 positive. This activation occurs as CD8+ T cells recognize the peptide, leading to an immune response. The NY-ESO-1 (157-165) peptide is present in various tumors, making it a potential target for tumor immunotherapy.
  • Peptide fragment from NY-ESO-1 protein.
  • Activates the immune system.
  • Recognized by CD8+ T cells in HLA-A2 positive individuals.
  • Triggers an immune response.
  • Expressed in various tumors.
  • Can be used as a target for tumor immunotherapy.
  • Highly immunogenic epitope of NY-ESO-1.
  • Can induce spontaneous or vaccine-induced T cell immune responses.
Catalog No. 50-004-23222