Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NY-ESO-1 (157-165) peptide | 202815-17-6 | 99.8% | 1094.35 | 5 MG
SDP

Supplier:  Medchemexpress LLC HYP104935MG

Encompass_Preferred

NY-ESO-1 (157-165) peptide is a peptide fragment derived from the NY-ESO-1 protein. It has the ability to activate the immune system. Specifically, in individuals who are positive for HLA-A2, it can be recognized by CD8+ T cells, thereby initiating an immune response. The NY-ESO-1 (157-165) peptide is expressed in various types of tumors, making it a potential target for tumor immunotherapy. It is considered a highly immunogenic epitope of NY-ESO-1 and can induce spontaneous or vaccine-induced T cell immune responses.

  • Purity: 99.8%
  • CAS number: 202815-17-6
  • Molecular weight: 1094.35
  • Formula: C49H79N11O13S2
  • Appearance: Solid
  • Color: White to off-white
  • Sequence: Ser-Leu-Leu-Met-Trp-Ile-Thr-Gln-Cys
  • Sequence shortening: SLLMWITQC

Catalog No. 50-004-25121


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.