Learn More
Medchemexpress LLC Ny-eso-1 (157-165) peptide Tfa | >97.6% | 1208.37 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP10493A5MG
Encompass_Preferred
NY-ESO-1 (157-165) peptide (TFA) is a peptide fragment derived from the NY-ESO-1 protein. It is capable of activating the immune system, particularly in HLA-A2 positive individuals, by being recognized by CD8+ T cells to initiate an immune response. This peptide is expressed in various tumors and is considered a potential target for tumor immunotherapy.
  • Highly immunogenic epitope of NY-ESO-1
  • Capable of inducing spontaneous or vaccine-induced T cell immune responses
  • Expressed in various tumors, making it a target for immunotherapy
Catalog No. 50-004-23231