Learn More
MSC PIP Boss Mechanic Gloves, Pigskin Leather, Impact Protection, 2XL, 1 Pair
Supplier:  MSC 13629175
Encompass
These mechanic gloves feature a premium top grain pigskin leather palm and a breathable mesh fabric back, designed to provide comfort and protection in various industrial applications. The porous pigskin construction offers breathability and maintains its soft texture even when exposed to moisture. Equipped with TPR impact protection on the fingers and back of the hand, these gloves are ideal for tasks requiring dexterity and impact resistance.
  • Top grain pigskin leather palm provides durability and softens with use
  • TPR impact protection on fingers and back of hand shields against impacts
  • Breathable mesh fabric back offers ventilation
  • Pre-curved fingers improve comfort and reduce hand fatigue
  • Integrated palm pad absorbs and dampens shock
  • Adjustable hook and loop closure ensures a secure fit
  • Four-way stretch thumb provides increased flexibility
Catalog No. 50-006-86883