Learn More
Sterlitech Corporation Nylon Membrane Filters, with Grid, 0.45 Micron, 47mm, 100/Pk
SDP
Supplier:  Sterlitech Corporation NG4547100
Encompass_Preferred
Nylon membrane disc filters: 0.45 µm pore-size, 47 mm diameter, pack of 100. Hydrophilic, low-extractable, inert, durable material supported by an internal polyester web. Typically used for HPLC, microbiology, medical/clinical, and general filtration.
, Gridded
Catalog No. NC1087494