Learn More
Tisch Scientific Tisch Scientific Nylon Membrane Filter, 1.0 µm, 8 x 10 in, Nonsterile, 25 Pack, SPEC20112
SDP
Supplier:  Tisch Scientific SPEC20112
Encompass
Tisch nylon membrane filters are naturally hydrophilic, have high non-specific binding properties, and an extractable level of <0.0015 mg/cm^2, they are ideal for HPLC solvent and sample preparation.
Catalog No. NC2090018