Learn More
Accuflow Systems NYLON TC BIO CLAMP 2.5IN 10/PK
Supplier:  Accuflow Systems NG250WHT
Encompass_Preferred
Reinforced nylon sanitary 2.5" Bio-Clamps, designed for use in GMP Manufacturing and laboratories. These clamps can be autoclavable to 275F as well as gama irradiated. Meet USP Class VI standards. Designed for Sanitary Tri clamp applications. Pack of 10
Catalog No. 50-110-8231