Learn More
Chemglass Life Sciences Nylon Tubing Clamp Kit
SDP
Supplier:  Chemglass Life Sciences CG15250
Encompass_Preferred
Nylon Clamp Kit (Part No. CG-152-50)A great way to be prepared for your next clamping situation!Kit contains 5 each of all 13 sizes of the CG152 tubing clamp series, neatly organized in a polypropylene box with adjustable dividers. Ideal for vacuum, pressure and liquid applications Chemical- and corrosion-resistant Can be sterilized for biological and medical applications Easy to install and remove by hand or with pliers Push down on one side to release
Catalog No. 50-131-1062