Learn More
Medchemexpress LLC Nystatin | 1400-61-9 | 95.7% | 926.09 | 1 G
SDP
Supplier:  Medchemexpress LLC HY174091G
Encompass_Preferred
Nystatin is an orally active polyene antifungal antibiotic effective against yeast and mycoplasma. It increases the permeability of plasma membranes to small monovalent ions, including chloridion. Nystatin is also a cholesterol-sequestering agent that partially prevents Oxaliplatin-induced lipid raft aggregation, DR4 and DR5 clustering, and thereby reduces apoptosis.
  • Orally active polyene antifungal antibiotic.
  • Effective against yeast and mycoplasma.
  • Increases permeability of plasma membranes to small monovalent ions, including chloridion.
  • Cholesterol-sequestering agent.
  • Partially prevents Oxaliplatin-induced lipid raft aggregation, DR4 and DR5 clustering, and reduces apoptosis.
Catalog No. 50-003-83966