Learn More
Crescent Chemical Co Inc NYSTATIN MIN. 4400 UNITS/MG
SDP
Supplier:  Crescent Chemical Co Inc SE29870.01
Encompass_Preferred
Nystatin min. 4 400 units/mg research grade, Ph. Eur. Polyene macrolide antibiotic isolated from Streptomyces noursei. Used in animal and plant cell cultures against fungi and yeasts. Forms complexes with ergosterol in the fungal cell membrane resulting in the formation of po
Catalog No. 50-247-890