Learn More
TOKU-E NYSTATIN SOLN 1ML
Supplier:  TOKU-E N0901ML
Encompass_Preferred
Antibiotic, ReadyMade™, sterile solution, 20,000-40,000 IU/ml in 0.02N HCl. Useful anti-mycotic used to control contamination in cell culture, plant biology and in susceptibility testing.
Catalog No. NC2068774