Learn More
Medchemexpress LLC Nystose | 13133-07-8 | 666.58 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN149910MM/1ML
Encompass_Preferred
Nystose is a tetrasaccharide composed of two fructose molecules linked via beta (1→2) bonds to the fructosyl moiety of sucrose. It exhibits prebiotic, immunomodulatory, and metabolism-regulating activities. Nystose promotes bone mineralization by activating the Wnt/β-catenin signaling pathway.
  • Exhibits prebiotic, immunomodulatory, and metabolism regulating activities.
  • Promotes bone mineralization by activating the Wnt/β-catenin signaling pathway.
  • Can be fermented by lactobacilli (L. intestinalis and L. reuteri) and promotes their proliferation.
  • Increases the activity of ALP and the proliferation of BMSCs.
  • Enhances intestinal IgA secretion and modulates systemic immune responses.
  • Improves bone density and microstructure in mouse models.
Catalog No. 50-004-15607