Learn More
Medchemexpress LLC Nystose | 13133-07-8 | 666.58 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN1499100MG
Encompass_Preferred
Nystose is a tetrasaccharide formed by two fructose molecules linked via beta (1→2) bonds to the fructosyl moiety of sucrose. It exhibits prebiotic, immunomodulatory, and metabolism-regulating activities, and also promotes bone mineralization.
  • Exhibits prebiotic, immunomodulatory, and metabolism-regulating activities.
  • Promotes bone mineralization by activating the Wnt/β-catenin signaling pathway.
  • Can be fermented by lactobacilli (L. intestinalis and L. reuteri).
  • Promotes the proliferation of lactobacilli.
  • Increases the activity of alkaline phosphatase (ALP).
  • Promotes the proliferation of bone marrow stromal cells (BMSCs).
Catalog No. 50-004-15674