Learn More
Medchemexpress LLC Nystose | 13133-07-8 | MFCD00191672 | 666.58 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN14995MG
Encompass_Preferred
Nystose is a tetrasaccharide with two fructose molecules linked via beta (1→2) bonds to the fructosyl moiety of sucrose. It exhibits prebiotic, immunomodulatory, and metabolism regulating activities.
  • Promotes bone mineralization by activating the Wnt/β-catenin signaling pathway.
  • Fermented by lactobacilli, promoting lactobacilli proliferation.
  • Enhances intestinal IgA secretion and modulates systemic immune responses.
  • Improves bone density and bone microstructure.
Catalog No. 50-004-14875