Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp NZM Broth 250 G
SDP

Supplier:  Research Products International Corp N71000250.0

Encompass_Preferred

Enriched medium for cultivating recombinant strains of E. Coli and propagating theta bacteriophage.

Dissolve 16 grams in 1 liter distilled water. Sterilize by autoclaving for 15 minutes.

Ingredients:

Casein Digest Protein: 10 g/L
Sodium Chloride: 5 g/L
Magnesium Sulfate: 0.94 g/L

Catalog No. 50-223-0446


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.