Learn More
Bioworld NZM Broth, 500 g
Supplier:  Bioworld 306210221
Encompass_Preferred
NZM BrothUsed for the cultivation of various genetic strains of E. coli Formulation recommended for replication of recombinant λ phageDoes not contain Casamino Acids or Yeast ExtractpH: 7.0Dehydrated Appearance: light beige, homogeneous powderPrepared Appearance: light to medium amber brothSoluble in waterComposition:Bacto Casein Digest Sodium Chloride Magnesium Sulfate
Catalog No. 50-254-444
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.