Learn More
Bioworld NZYM Broth, 500 g
Supplier:  Bioworld 306210231
Encompass_Preferred
NZYM BrothUsed for the cultivation of various genetic strains of E. coli Formulation recommended for replication of recombinant λ phageDoes not contain Casamino AcidspH: 7.0Dehydrated Appearance: light beige, homogeneous powderPrepared Appearance: light to medium amber brothSoluble in waterComposition:Bacto Casein DigestBacto Yeast ExtractSodium ChlorideMagnesium Sulfate
Catalog No. 50-254-445
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.