Learn More
Medchemexpress LLC o-3M3FBS | 313981-55-4 | 343.36 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY108608
Encompass_Preferred
o-3M3FBS is the negative control of m-3M3FBS. It inhibits inward and outward currents through mechanisms independent of PLC, acting antagonistically. Additionally, o-3M3FBS can increase [Ca2+]i agonistically. In a study, o-3M3FBS (10 μM) decreased AUC from 1058±189 g/min in control (with TTX) to 840±134 g/min, and a higher concentration (25 μM) further reduced AUC to 457±59 g/min.
  • Negative control of m-3M3FBS.
  • Inhibits inward and outward currents via PLC-independent mechanisms.
  • Acts antagonistically.
  • Increases [Ca2+]i agonistically.
  • Suitable for research on calcium channel activity and neuronal signaling.
  • Soluble in various solvents for in vitro and in vivo studies.
Catalog No. 50-003-47488