Learn More
Selleck Chemical LLC o-3M3FBS-E0778-5MG
SDP
Supplier:  Selleck Chemical LLC E07785MG
Encompass_Preferred
O-3M3FBS the negative control of m-3M3FBS inhibits inward and outward currents via mechanisms independent of PLC acting in an antagonistic manner reversibly inhibits delayed rectifier K channels increases in [Ca2 ](i) in an agonistic manner with m-3M3FBS
Catalog No. 50-313-5790