Learn More
Medchemexpress LLC O-acetyl-L-serine | 5147-00-2 | MFCD00037771 | >=98.0% | 147.13 g/mol | C5H9NO4 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY101409R25MG
Encompass_Preferred
O-Acetylserine (O-acetyl-L-serine) is an endogenous metabolite and an analytical standard used in research to study sulfur assimilation and cysteine biosynthesis. It functions as a biochemical intermediate and signaling molecule that can influence expression of sulfate transporters and adenosine 5'-phosphosulfate reductase. Supplied for analytical and research applications in small, quantified amounts.
  • Analytical standard suitable for biochemical and analytical research.
  • Endogenous metabolite involved in cysteine biosynthesis.
  • Modulates gene expression of sulfate transporters and APS reductase.
  • Molecular formula C5H9NO4 and molecular weight 147.13 g/mol.
  • Supplied in small, quantified mg quantities for analytical use.
Catalog No. 50-002-45413