Learn More
Cayman Chemical O-desethyl DapaglIflozIn 50mg
SDP
Supplier:  Cayman Chemical 3602250 MG
Encompass_Preferred
An SGLT2 inhibitor (IC50 = 9.4 nM) and active metabolite of dapagliflozin; selective for SGLT2 over SGLT1 (IC50 = 970 nM)
Catalog No. 50-290-9191