Learn More
Medchemexpress LLC HY-N4075 5mg Medchemexpress, O-Desmethylangolensin CAS:21255-69-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN40755MG
Encompass_Preferred
Medchemexpress, HY-N4075 5mg O-Desmethylangolensin CAS:21255-69-6 O-Desmethylangolensin is a metabolite of soy isoflavone, daidzein metabolized by gut microbiota. O-Desmethylangolensin possesses antioxidant activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2920