Learn More
Medchemexpress LLC O-Methyldauricine | 2202-17-7 | 1 MG

Supplier: Medchemexpress LLC HYN26121MG
O-Methyldauricine is an alkaloid separated from P. ophthalmicus, known for its antibacterial activity. This product has a purity of 96.08% and is classified as a piperidine alkaloid.
- Alkaloid with a benzylisoquinoline group
- Exhibits antibacterial activity
- Molecular weight: 638.79
- Chemical formula: C39H46N2O6
- Appearance: Solid
- Color: Yellow to brown
- Initial source: Plants, Menispermaceae, Hypserpa nitida Miers
- Related to infection and anti-infection disease areas
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.