Learn More
Medchemexpress LLC O-Methyldauricine | 2202-17-7 | 5 MG

Supplier: Medchemexpress LLC HYN26125MG
O-Methyldauricine is an alkaloid with a benzylisoquinoline group, separated from P. ophthalmicus. It exhibits antibacterial activity and has a purity of 96.08%.
- Molecular formula: C39H46N2O6
- Molecular weight: 638.79
- Appearance: Solid
- Color: Yellow to brown
- Structure classification: Alkaloids, piperidine alkaloids
- Initial source: Plants, Menispermaceae, Hypserpa nitida Miers
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.