Learn More
Apexbio Technology LLC O-Phospho-L-serine 407-41-0 100mg
SDP
Supplier:  Apexbio Technology LLC B6233100
Encompass_Preferred
O-Phospho-L-serine (CAS 407-41-0) is a metabolic intermediate in the glutamate pathway and plays a role in the biosynthesis of L-cysteine It exhibits selective modulation of metabotropic glutamate receptors (mGluRs) functioning as an agonist at group III subtypes (mGluR4 mGluR6 mGluR7 mGluR8) while acting as an antagonist at selected receptors within groups I and II (notably mGluR1 and mGluR2) This profile enables O-Phospho-L-serine to serve as a tool compound for investigating glutamate-mediated neuronal signaling pathways and assessing pharmacological effects associated with mGluR modulation in cellular and neurobiological research contexts
Catalog No. 50-000-89648