Learn More
Medchemexpress LLC O-Phospho-L-serine | 407-41-0 | C3H8NO6P | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY15129R
Encompass_Preferred
O-Phospho-L-serine is an endogenous metabolite that acts as the immediate precursor to L-cystein in the serine synthesis pathway. It functions as an agonist at group III mGluR receptors (mGluR4, mGluR6, mGluR7, and mGluR8), while also serving as a weak antagonist for mGluR1 and a potent antagonist for mGluR2. This product is intended for research use only.
  • Weakly binds to mGluR1 and antagonizes the effects of l-glutamate
  • Activates group III receptors (mGluR4, mGluR6, mGluR7, and mGluR8)
  • Enhances intracellular calcium responses in mGluR4 transfected cells
  • Inhibits l-glutamate mediated mGluR1 response
  • Potently inhibits mGluR2 activation
  • Induces membrane potential changes in HEK/TRPC4 cells transfected with mGluR4 or mGluR6, mediating TRPC4β activation
  • Inhibits Müller glia proliferation in light-damaged retinas
  • Inhibits cone cell regeneration in light-damaged retinas
Catalog No. 50-003-76000