Learn More
Selleck Chemical LLC O-Phospho-L-serine S5137-1g
SDP
Supplier:  Selleck Chemical LLC S51371G
Encompass_Preferred
O-Phospho-L-serine (Dexfosfoserine) a molecule that mimics the phosphatidylserine head group and partially blocks microglial phagocytosis of apoptotic cells is a specific group III metabotropic glutamate receptor (mGluR) agonist
Catalog No. 50-311-2318