Learn More
Cayman Chemical O-Phospho-L-TyrosIn 500mg
SDP
Supplier:  Cayman Chemical 23064500 MG
Encompass_Preferred
A phosphorylated form of L-tyrosine; used in studies of tyrosine phosphorylation; has been used in affinity chromatography to purify IgG
Catalog No. 50-299-5575