Learn More
Cayman Chemical O-PhosphoethnolamIn 100g
SDP
Supplier:  Cayman Chemical 34636100 G
Encompass_Preferred
A phospholipid metabolite; precursor in the biosynthesis of phosphatidylethanolamines in the endoplasmic reticulum; decreases disease severity in a mouse model of cerulein-induced acute pancreatitis; O-phosphoethanolamine levels are decreased in postmortem amygdala from patients with Parkinson's disease
Catalog No. 50-295-1007