Learn More
Medchemexpress LLC O-proparagyl-N-boc-ethanolamine | 634926-63-9 | MFCD22418470 | >95.0% | 199.25 | C10H17NO3 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW2595421G
Encompass_Preferred
O-Proparagyl-N-Boc-ethanolamine is a Boc-protected propargyl ethanolamine used as a click-chemistry crosslinker and synthetic building block for bioconjugation and small-molecule synthesis. It features a terminal alkyne for azide coupling and a t-Boc protected amine that can be removed under mild acidic conditions. For research use only.
Catalog No. 50-002-81528