Learn More
Medchemexpress LLC O-proparagyl-N-Boc-ethanolamine | 634926-63-9 | MFCD22418470 | 95.0% | 199.25 g/mol | C10H17NO3 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW259542500MG
Encompass_Preferred
O-Proparagyl-N-Boc-ethanolamine is a Boc-protected ethanolamine bearing a propargyl (alkyne) functional group. It serves as a synthetic building block and alkyne handle for organic synthesis, enabling conjugation via click chemistry and incorporation as a linker or crosslinker in medicinal chemistry applications. Supplied as a solid reagent for routine laboratory synthesis and derivatization.
Catalog No. 50-002-81208