Learn More
Medchemexpress LLC O-proparagyl-n-boc-ethanolamine | 634926-63-9 | MFCD22418470 | >97.0% | 199.25 | C10H17NO3 | 250 MG
SDP
Supplier:  Medchemexpress LLC HYW259542250MG
Encompass_Preferred
O-Proparagyl-N-Boc-ethanolamine is a Boc-protected ethanolamine with a propargyl (terminal alkyne) functional group designed for copper-catalyzed azide-alkyne cycloaddition (click chemistry). The Boc group protects the amine during reactions and can be removed under mild acidic conditions to expose a reactive primary amine for subsequent modifications.
  • Enables CuAAC-based bioconjugation via a terminal alkyne.
  • Contains a Boc-protected amine that is removable under mild acidic conditions.
  • Functions as a small-molecule crosslinker for labeling or tethering biomolecules.
  • Compatible with common organic solvents and standard synthetic workflows.
  • Provides defined molecular weight and formula for analytical tracking.
Catalog No. 50-002-81213