Learn More
Selleck Chemical LLC O-Propargyl-Puromycin E4666-25MG
SDP
Supplier:  Selleck Chemical LLC E466625MG
Encompass_Preferred
O-Propargyl-Puromycin (OP-puro) is an alkyne puroanalog a potent inhibitor of protein synthesis It is used to label nascent proteins in whole animals allowing the visualization of protein synthesis patterns in large explants from tissues and organs
Catalog No. 50-000-39008