Learn More
Ace Glass, Inc. O-RING CAPFE SIZE 013 O-RING CAPFE SIZE 013
SDP
Supplier:  Ace Glass, Inc. 7855810
Encompass_Preferred
Size 013 CAPFE o-ring is an ACE-exclusive o-ring with a resilient rubber core encased in a thick, non-porous FEP/PTFE encapsulation. CAPFE has excellent compression set and chemical compatibility, and low gas and water permeability.
Catalog No. 50-975-711